2,3-Dimethoxy-5-methyl-p-benzoquinone 98%

Supplier: Thermo Fisher Scientific
Warning

442200010 442200050
ACRO442200010EA 55.5 EUR
ACRO442200010 ACRO442200050
2,3-Dimethoxy-5-methyl-p-benzoquinone 98%
2,3-Dimethoxy-5-methyl-p-benzoquinone
Formula: C₉H₁₀O₄
Melting Pt: 59…61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001595
CAS Number: 605-94-7

Order Now

Specification Test Results

Appearance Orange to red Crystalline powder or needles
Infrared spectrum Conforms
Melting point 58 to 62 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR