N-Boc-1,3-propanediamine 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: N-(3-Aminopropyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(3-Aminopropyl)carbamate, N-Boc-1,3-propanediamine, tert-Butyl (3-aminopropyl)carbamate, N-1-Boc-1,3-diaminopropane·HCl, N-(tert-Butoxycarbonyl)-1,3-propanediamine, N-Boc-1,3-diaminopropane

442730010 442730050
ACRO442730010EA 96 EUR
ACRO442730010 ACRO442730050
N-Boc-1,3-propanediamine 97%
N-Boc-1,3-propanediamine
Formula: C₈H₁₈N₂O₂
MW: 174.244 g/mol
Boiling Pt: 94…96 °C (.2Torr)
Melting Pt: 22 °C
Density: 0.998 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00210021
CAS Number: 75178-96-0
UN: 3259
ADR: 8 III

Order Now

Specification Test Results

Appearance Colorless to white Liquid or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR