4-Chlorophenylboronic acid pinacol ester 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-(4-Chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Chlorophenylboronic acid Pinacol ester, 4-Chlorobenzeneboronic acid pinacol ester, 1-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

444690050 444690250
ACRO444690050EA 81.2 EUR
ACRO444690050 ACRO444690250
4-Chlorophenylboronic acid pinacol ester 97%
4-Chlorophenylboronic acid pinacol ester
Formula: C₁₂H₁₆BClO₂
MW: 238.52 g/mol
Melting Pt: 55…56 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD05663875
CAS Number: 195062-61-4

Order Now

Specification Test Results

Appearance White to off-white Solid or crystalline solid
Infrared spectrum Conforms
Melting point 54 to 59 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR