4-(Methylsulphonylamino)benzeneboronic acid 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-(Methylsulphonylamino)phenylboronic acid, 4-Methylsulphonylaminophenylboronic acid, 4-(Methanesulphonamido)phenylboronic acid

445470010 445470050
ACRO445470010EA 111 EUR
ACRO445470010 ACRO445470050
4-(Methylsulphonylamino)benzeneboronic acid 97%
4-(Methylsulphonylamino)benzeneboronic acid
Formula: C₇H₁₀BNO₄S
MW: 215.03 g/mol
Melting Pt: 234…236 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179473
CAS Number: 380430-57-9

Order Now

Specification Test Results

Appearance Off-white to cream to light brown Solid
Infrared spectrum Conforms
Additional info May contain varying amounts of anhydrides
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR