N-Methyl-2-pyrrolidone (NMP) 99.5% for analysis

Supplier: Thermo Fisher Scientific
Danger

445790010 445790025
ACRO445790010EA 142 EUR
ACRO445790010 ACRO445790025
N-Methyl-2-pyrrolidone (NMP) 99.5% for analysis
N-Methyl-2-pyrrolidone (NMP)
Formula: C₅H₉NO
Boiling Pt: 202 °C (760 mmHg)
Melting Pt: –24 °C
Density: 1,03 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.1 %
GC ≥99.4 %
Color scale ≤50 APHA
Residue after ignition ≤0.01 %
Aluminium (Al) ≤1 ppm
Antimony (Sb) ≤1 ppm
Arsenic (As) ≤1 ppm
Boron (B) ≤1 ppm
Calcium (Ca) ≤5 ppm
Chromium (Cr) ≤1 ppm
Copper (Cu) ≤1 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤1 ppm
Magnesium (Mg) ≤1 ppm
Manganese (Mn) ≤1 ppm
Nickel (Ni) ≤1 ppm
Potassium (K) ≤20 ppm
Sodium (Na) ≤20 ppm
Zinc (Zn) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR