Benzyl alcohol ACS

Supplier: Thermo Fisher Scientific
Warning

447720010 447720025 447721000
ACRO447720010EA 160 EUR
ACRO447720010 ACRO447720025 ACRO447721000
Benzyl alcohol ACS
Benzyl alcohol
Formula: C₆H₅CH₂OH
Boiling Pt: 205 °C (760 mmHg)
Melting Pt: –15 °C
Density: 1,044 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
GC ≤0.01 % Benzaldehyde
Color scale ≤20 APHA
Ash ≤0.005 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR