Ammonium molybdate(VI) 99.99% (trace metals basis)

Supplier: Thermo Fisher Scientific
Warning

448540050 448540250 448541000
ACRO448540050EA 150 EUR
ACRO448540050 ACRO448540250 ACRO448541000
Ammonium molybdate(VI) 99.99% (trace metals basis)
Ammonium molybdate(VI)
Formula: H₈MoN₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00011596
CAS Number: 13106-76-8

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals and/or chunks
Total trace metal impurities ≤100 ppm
Molybdenum (Mo) 47.0 to 50.7 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR