5-Amino-3-methyl-1-phenyl-1H-pyrazole 97%

Supplier: Thermo Fisher Scientific
Warning

449600050 449600250
ACRO449600050EA 56 EUR
ACRO449600050 ACRO449600250
5-Amino-3-methyl-1-phenyl-1H-pyrazole 97%
5-Amino-3-methyl-1-phenyl-1H-pyrazole
Formula: C₁₀H₁₁N₃
Boiling Pt: 333 °C (760 mmHg)
Melting Pt: 113…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00020727
CAS Number: 1131-18-6

Order Now

Specification Test Results

Appearance White to brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 114 to 117 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR