4'-Fluorobiphenyl-4-sulphonyl chloride 97%

Supplier: Thermo Fisher Scientific
Danger

450320050
ACRO450320050EA 173 EUR
ACRO450320050
4'-Fluorobiphenyl-4-sulphonyl chloride 97%
4'-Fluorobiphenyl-4-sulphonyl chloride
Formula: C₁₂H₈ClFO₂S
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD01631921
CAS Number: 116748-66-4
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 80 to 85 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR