Ammonium bromide 98%, extra pure

Supplier: Thermo Fisher Scientific
Warning

450650010 450650025 450650100
ACRO450650010EA 59.7 EUR
ACRO450650010 ACRO450650025 ACRO450650100
Ammonium bromide 98%, extra pure
Ammonium bromide
Formula: H₄BrN
Boiling Pt: 235 °C (760 mmHg)
Melting Pt: 452 °C (subl.)
Density: 2,42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011419
CAS Number: 12124-97-9

Order Now

Specification Test Results

Appearance White Crystalline powder and/or chunks
Titration Argentometric ≥97.5 %
Sulfated ash ≤0.1 %
Insoluble matter ≤0.01 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR