Sulphuric acid 25% water analysis_for

Supplier: Thermo Fisher Scientific
Danger

451050010
ACRO451050010EA 62.5 EUR
ACRO451050010
Sulphuric acid 25% water analysis_for
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 2796
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear liquid
Color scale ≤10 APHA
Residue after ignition ≤0.0005 %
Titration with NaOH ≥25 %
Chloride (Cl) ≤0.5 ppm
Nitrate (NO3) ≤0.2 ppm
Aluminium (Al) ≤0.05 ppm
Ammonium (NH4) ≤2 ppm
Arsenic (As) ≤0.01 ppm
Beryllium (Be) ≤0.02 ppm
Cadmium (Cd) ≤0.02 ppm
Calcium (Ca) ≤0.2 ppm
Chromium (Cr) ≤0.05 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.01 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.02 ppm
Lithium (Li) ≤0.02 ppm
Magnesium (Mg) ≤0.05 ppm
Manganese (Mn) ≤0.01 ppm
Nickel (Ni) ≤0.02 ppm
Potassium (K) ≤0.1 ppm
Silver (Ag) ≤0.02 ppm
Sodium (Na) ≤0.5 ppm
Strontium (Sr) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm
Heavy metals (as Pb) ≤1 ppm
KMnO4-reducing substance ≤0.0002 % (as SO2)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR