Perchloric acid ∼70% water, extra_pure

Supplier: Thermo Fisher Scientific
Danger

451770025
ACRO451770025EA 242 EUR
ACRO451770025
Perchloric acid ∼70% water, extra_pure
Perchloric acid
Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 203 °C (760 mmHg)
Melting Pt: –18 °C
Density: 1.66 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
UN: 1873
ADR: 5.1 I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear solution
Color scale ≤10 APHA
Residue after ignition ≤0.003 %
Titration with NaOH 70 to 72 %
Chloride (Cl) ≤3 ppm
Sulfate (SO4) ≤5 ppm
Iron (Fe) ≤1 ppm
Heavy metals (as Pb) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR