Cadmium(II) bromide, anhydrous 98%, pure

Supplier: Thermo Fisher Scientific
Warning

453860250 453861000 453865000
ACRO453860250EA 59.8 EUR
ACRO453860250 ACRO453861000 ACRO453865000
Cadmium(II) bromide, anhydrous 98%, pure
Cadmium (II) bromide
Formula: Br₂Cd
Boiling Pt: 963 °C
Melting Pt: 566 °C
Density: 5,19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00015996
CAS Number: 7789-42-6
UN: 2570
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Assay ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR