2,2,2-Trifluoroethyl trifluoromethanesulphonate 97%

Supplier: Thermo Fisher Scientific
Danger

454970010 454970050 454970250
ACRO454970010EA 55 EUR
ACRO454970010 ACRO454970050 ACRO454970250
2,2,2-Trifluoroethyl trifluoromethanesulphonate 97%
2,2,2-Trifluoroethyl trifluoromethanesulphonate
Formula: C₃H₂F₆O₃S
Boiling Pt: 89…91 °C (740 mmHg)
Density: 1,61 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00671579
CAS Number: 6226-25-1
UN: 2927
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR