Penicillin V potassium salt 99%

Supplier: Thermo Fisher Scientific
Danger

455230250 455230050
ACRO455230250EA 132 EUR
ACRO455230250 ACRO455230050
Penicillin V potassium salt 99%
Penicillin V potassium salt
Formula: C₁₆H₁₇KN₂O₅S
Melting Pt: 197…202 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051771
CAS Number: 132-98-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Specific optical rotation +220° to +235° (20°C, 589 nm) (c=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR