Amphotericin B 85%

Supplier: Thermo Fisher Scientific
Warning

455490010 455490050
ACRO455490010EA 160 EUR
ACRO455490010 ACRO455490050
Amphotericin B 85%
Amphotericin B
Formula: C₄₇H₇₃NO₁₇
Melting Pt: >170 °C
Storage Temperature: Freezer
MDL Number: MFCD00877763
CAS Number: 1397-89-3

Order Now

Specification Test Results

Appearance Yellow to orange Powder
UV spectrum Complies
Water ≤10 %
HPLC ≥80 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR