Rosuvastatin calcium salt 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 7-{4-(4-Fluorophenyl)-6-isopropyl-2-[methyl(methylsulphonyl)amino]pyrimidin-5-yl}-3,5-dihydroxyhept-6-enoic acid, Calcium (3R,5S,E)-7-(4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethylsulphonamido)pyrimidin-5-yl)-3,5-dihydroxyhept-6-enoate, Rosuvastatin Calcium, CRESTOR, (3R,5S,6E)-7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[N-methyl(n-methylsulphonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-6-Heptenoic calcium

456230010 456232500
ACRO456230010EA 378 EUR
ACRO456230010 ACRO456232500
Rosuvastatin calcium salt 98%
Rosuvastatin calcium salt
Formula: C₂₂H₂₇FN₃O₆S·¹/₂Ca
MW: 501.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04112702
CAS Number: 147098-20-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR