Ondansetron hydrochloride dihydrate 98%

Supplier: Thermo Fisher Scientific
Danger

461160010 461160050
ACRO461160010EA 72.8 EUR
ACRO461160010 ACRO461160050
Ondansetron hydrochloride dihydrate 98%
Ondansetron hydrochloride dihydrate
Formula: C₁₈H₁₉N₃O·HCl·₂H₂O
Melting Pt: 178 °C
Storage Temperature: Freezer
MDL Number: MFCD00374371
CAS Number: 103639-04-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 176 to 180 °C
Water ≤10.5 %
HPLC ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR