Procarbazine hydrochloride

Supplier: Thermo Fisher Scientific
Danger

461180010 461182500
ACRO461180010EA 570 EUR
ACRO461180010 ACRO461182500
Procarbazine hydrochloride
Procarbazine hydrochloride
Formula: C₁₂H₁₉N₃O·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00072082
CAS Number: 366-70-1

Order Now

Specification Test Results

Appearance White to tan Powder
Infrared spectrum Conforms
Loss on drying ≤2 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR