Sulphasalazine 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 5-[4-(2-Pyridylsulphamoyl)phenylazo]salicylic Acid, 2-Hydroxy-5-((4-(N-(pyridin-2-yl)sulphamoyl)phenyl)diazenyl)benzoic acid

461240050 461240250 461241000
ACRO461240050EA 48.5 EUR
ACRO461240050 ACRO461240250 ACRO461241000
Sulphasalazine 97%
Sulphasalazine
Formula: C₁₈H₁₄N₄O₅S
MW: 398.39 g/mol
Melting Pt: 260…265 °C
Storage Temperature: Ambient
MDL Number: MFCD00057363
CAS Number: 599-79-1

Order Now

Specification Test Results

Appearance Light yellow to brownish-yellow Powder
Infrared spectrum Conforms
Loss on drying ≤1 % (1 g, 105°C, 2 h)
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR