Rosiglitazone maleate

Supplier: Thermo Fisher Scientific

462410010
ACRO462410010EA 160 EUR
ACRO462410010
Rosiglitazone maleate
Rosiglitazone maleate

  • Bioactive

Formula: C₁₈H₁₉N₃O₃S·C₄H₄O₄
Melting Pt: 123 °C
Storage Temperature: Ambient
MDL Number: MFCD03427306
CAS Number: 155141-29-0

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 118 to 123 °C
Loss on drying ≤0.5 %
Titration with HClO4 98 to 102 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR