Paroxetine hydrochloride hemihydrate ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (3S,4R)-(-)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine Hydrochloride

462630010 462630050
ACRO462630010EA 99.4 EUR
ACRO462630010 ACRO462630050
Paroxetine hydrochloride hemihydrate ≥98%
Paroxetine hydrochloride hemihydrate

  • Bioactive

Formula: C₁₉H₂₀FNO₃
MW: 365.83 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03658863
CAS Number: 110429-35-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water 2.0 to 3.0 %
Solubility (5 % in methanol) Clear colorless solution
HPLC ≥97.5 %
Residue after ignition ≤0.1 % (Platinum crucible)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR