Calcium sulphate, Drierite™ 6 mesh, with indicator

Supplier: Thermo Fisher Scientific
Danger

463475000 463470020
ACRO463475000EA 109 EUR
ACRO463475000 ACRO463470020
Calcium sulphate, Drierite™ 6 mesh, with indicator
Calcium sulphate
Formula: CaO₄S
Melting Pt: 1450 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010912
CAS Number: 7778-18-9
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Blue Irregular granules
Water ≤0.3 %
Particle size 6 mesh
Composition ≤2 % (CoCl2) Typical


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR