Ethanol denatured (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Ethyl alcohol denatured

463680010 463680025 463680050 463680250
ACRO463680010EA 50 EUR
ACRO463680010 ACRO463680025 ACRO463680050 ACRO463680250
Ethanol denatured (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Ethanol denatured

  • solvent

Formula: H₃CCH₂OH
MW: 46.07 g/mol
Boiling Pt: 78 °C (760 mmHg)
Melting Pt: –114.1 °C
Density: 0.79
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Refractive index 1.3610 to 1.3630 (20°C, 589 nm)
Water ≤3400 ppm
Density (20°C) 0.789 to 0.790 kg/l
Additional info Denatured with 1% iPr-OH, 1% MEK and 0.001% Bitrex
E/E 99.8 to 100.0 % v/v (calculated)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR