Magnesium(II) gluconate hydrate

Supplier: Thermo Fisher Scientific

Synonyms: D-Gluconic acid hemimagnesium salt hydrate, Magnesium D-gluconate hydrate, Gluconic acid magnesium(II) salt hydrate, D-Gluconic acid magnesium salt hydrate

464830250 464831000
ACRO464830250EA 41.5 EUR
ACRO464830250 ACRO464831000
Magnesium(II) gluconate hydrate
Magnesium(II) gluconate hydrate

  • Bioactive

Formula: C₆H₁₂O₇·xH₂O
Melting Pt: 169,7…171.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00150971
CAS Number: 3632-91-5

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Titration Complexometric 98.0 to 102.0 % (on dry substance)
Water ≤13.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR