Almotriptan Malate

Supplier: Thermo Fisher Scientific

Synonyms: 3-[2-(Dimethylamino)ethyl]-5-(pyrrolidin-1-ylsulphonylmethyl)-1H-indole Malate

465402500
ACRO465402500EA 495 EUR
ACRO465402500
Almotriptan Malate
Almotriptan Malate

  • Bioactive

Formula: C₁₇H₂₅N₃O₂S·C₄H₆O₅
Storage Temperature: Freezer
MDL Number: MFCD08067740
CAS Number: 181183-52-8

Order Now

Specification Test Results

Appearance to light orange Powder
Infrared spectrum Conforms
Water ≤0.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR