BIX 01294 95%

Supplier: Thermo Fisher Scientific
Warning

467831000
ACRO467831000EA 263 EUR
ACRO467831000
BIX 01294 95%
BIX 01294

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formula: C₂₈H₃₈N₆O₂
Storage Temperature: Freezer
MDL Number: MFCD14560563
CAS Number: 935693-62-2

Order Now

Specification Test Results

Appearance White to yellow to light brown Powder or crystalline powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR