GSK-583 95%

Supplier: Thermo Fisher Scientific
Warning

468211000
ACRO468211000EA 389 EUR
ACRO468211000
GSK-583 95%
GSK-583

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formula: C₂₀H₁₉FN₄O₂S
Storage Temperature: Freezer
MDL Number: MFCD30343846
CAS Number: 1346547-00-9

Order Now

Specification Test Results

Appearance White to yellow Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR