Zirconium standard solution, 1000 mg/L Zr in 5% hydrochloric acid with hydrofluoric acid (max. 1%) standard for ICP
Supplier: CPACHEM
C070.5C05FP.L1 C070.5C05FPL25 C070.5C05FP.L5
ACSDC070.5C05FP.L1EA 105 EUR
ACSDC070.5C05FP.L1 ACSDC070.5C05FPL25 ACSDC070.5C05FP.L5
Zirconium standard solution, 1000 mg/L Zr in 5% hydrochloric acid with hydrofluoric acid (max. 1%) standard for ICP
Standards Cationic Standards Zirconium Standards