Phosphate standard solution, 1,000 mg/L PO₄³⁻ in Water standard for ion chromatography
Supplier: CPACHEM
H045.W.L1 H045.W.L25 H045.W.L5
ACSDH045.W.L1EA 0 EUR
ACSDH045.W.L1 ACSDH045.W.L25 ACSDH045.W.L5
Phosphate standard solution, 1,000 mg/L PO₄³⁻ in Water standard for ion chromatography
Standards Anionic Standards Phosphate Standards