Bravo Disposable Pipette Tips
Supplier: AGILENT
19477-022 10734-202 10734-212 10734-302 21740-202 10734-312 21740-212 11484-202 11484-212 11484-302 21750-202 11484-312 21750-212 11484-322 19133-102 19133-112 19133-202 19133-212 19133-142 19134-012 19134-002 19134-022 19477-002 19477-012 19477-032 19477-072 19477-082
AGLS19477-022EA 1050 EUR
AGLS19477-022 AGLS10734-202 AGLS10734-212 AGLS10734-302 AGLS21740-202 AGLS10734-312 AGLS21740-212 AGLS11484-202 AGLS11484-212 AGLS11484-302 AGLS21750-202 AGLS11484-312 AGLS21750-212 AGLS11484-322 AGLS19133-102 AGLS19133-112 AGLS19133-202 AGLS19133-212 AGLS19133-142 AGLS19134-012 AGLS19134-002 AGLS19134-022 AGLS19477-002 AGLS19477-012 AGLS19477-032 AGLS19477-072 AGLS19477-082
Bravo Disposable Pipette Tips
Pipette Tips
Certified on Bravo and AssayMAP for accuracy and precision.
  • Individual lot certified to meet thorough QC standards
  • Certified free of detectable DNase and RNase activity
  • Certified free of endotoxin (pyrogen) contamination
  • Certified free of adenosine triphosphate (ATP)
  • Certified free of human DNA;Certified free of protease activity
  • Certified free of heavy metals
  • Certified as sterile with a sterility assurance level (SAL) of 10⁻⁶
  • Innovative processing and packaging reduce static buildup and improve deck space utilisation
Agilent offers disposable pipetteMore Product Information