AdEasy viral titer kit
Supplier: AGILENT
972500
AGLS972500EA 579 EUR
AGLS972500
AdEasy viral titer kit
Assays Immunological Assays
The AdEasy Viral Titer Kit is a simple enzyme-linked immunoassay for the determination of adenoviral titers.
The immunoassay detects an adenoviral capsid protein called hexon, which is required for adenoviral replication. The AdEasy viral titer kit can determine the titer of your stocks in 24 to 48 hours, compared to two weeks for traditional endpoint dilution assays.
Caution: For Research Use Only. Not for Use in Diagnostic Procedures.