Anti-kappa light chain Goat Polyclonal Antibody (DyLight® 650)

Supplier: Agrisera
AS122294
AGRIAS122294EA 282 EUR
AGRIAS122294
Anti-kappa light chain Goat Polyclonal Antibody (DyLight® 650)
Antibodies
Type: Secondary
Antigen: kappa light chain
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR