Pastette® transfer pipettes, standard
Supplier: Alpha Laboratories
LW4237 LW4021
ALPCLW4237EA 141 EUR
ALPCLW4237 ALPCLW4021
Pastette® transfer pipettes, standard
Pipettes Pasteur/Transfer Pipettes
Use these disposable Pastettes instead of a Pasteur pipette for liquid transfers.
Ideal for accessing small diameter tubes such as cuvettes and analyser cups. The stems can be heat-sealed to create a deep freeze, transport or storage container.
Designed to replace glass Pasteur pipettes, Pastette (transfer pipette) s are unbreakable and don't require a separate bulb. and, as they are manufactured from soft low density polyethylene (LDPE), can be recycled if used with non hazardous samples.
Packaging: Pack of 500 individually wrapped.
Order Now
SPECIFICATIONS
Environmentally Preferable