Anti-TBK1 (108A429) Mouse Monoclonal Antibody [clone: 108A429]

Supplier: Merck Millipore (Oncogene)
AM76-100
AM76-100EA 777 EUR
AM76-100
Anti-TBK1 (108A429) Mouse Monoclonal Antibody [clone: 108A429]
Antibodies
Type: Primary
Antigen: TBK1 (108A429)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR