Sleeve protectors, AlphaTec® 2000, model 600
Supplier: Ansell
WH20B-00600-00
ANSEWH20B-00600-00EA 686 EUR
ANSEWH20B-00600-00
Sleeve protectors, AlphaTec® 2000, model 600
Disposable Garments
These sleeve protectors are made from a non woven fabric with microporous PE film. The fabric provides an effective liquid and particle barrier combined with a high water vapour transmission rate from inside to outside. Ideal for a wide range of industrial applications.
  • Elasticated at both ends
  • Bound seams
  • Lightweight and comfortable
  • Excellent liquid penetration resistance and barrier to fine particulates
  • Silicone free: Critical in paint spraying applications
  • Anti-static: Tested according to EN 1149-5
Typical applications include: PharmaceuticalMore Product Information