Apron with sleeves, MICROCHEM® 3000, model 215
Supplier: Ansell
YE30W-00215-02
ANSEYE30W-00215-02EA 0 EUR
ANSEYE30W-00215-02
Apron with sleeves, MICROCHEM® 3000, model 215
Disposable Garments
This apron is made from an external film barrier layer with two non woven layers underneath. The fabric provides a strong liquid and particle barrier.
  • Velcro fastening to neck
  • Tie fastening at the waist
  • Double cuff with knitted inner cuff
  • Welded and taped seams
  • Multi-layer barrier fabric is effective against numerous chemicals
  • Highly visible: Bright yellow for improved worker safety
  • Lightweight yet durable
Typical applications include: Chemicals, oil and petrochemicals, pharmaceutical, food industry (caustic clean downs), sewage purificationMore Product Information