Anti-MRPS35 Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM
A100487-100
ANTIA100487-100EA 475 EUR
ANTIA100487-100
Anti-MRPS35 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to MRPS35 for WB and ELISA with samples derived from human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000

Type: Primary
Antigen: MRPS35
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR