Anti-Perforin Mouse Monoclonal Antibody [clone: PRF1/2468]

Supplier: ANTIBODIES.COM
A252892-100
ANTIA252892-100EA 520 EUR
ANTIA252892-100
Anti-Perforin Mouse Monoclonal Antibody [clone: PRF1/2468]
Antibodies
Mouse monoclonal [PRF1/2468] antibody to Perforin for ELISA with samples derived from Human.

  • Validated applications: ELISA

Type: Primary
Antigen: Perforin
Clonality: Monoclonal
Clone: PRF1/2468
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
New Product
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR