Anti-Lambda Light Chain Rabbit Recombinant Antibody [Clone: LLC/3774R]

Supplier: ANTIBODIES.COM
A278589-100
ANTIA278589-100EA 520 EUR
ANTIA278589-100
Anti-Lambda Light Chain Rabbit Recombinant Antibody [Clone: LLC/3774R]
Antibodies
Recombinant rabbit monoclonal [LLC/3774R] antibody to Lambda Light Chain for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Lambda Light Chain
Clonality: Recombinant
Clone: LLC/3774R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR