Anti-Sodium Iodide Symporter Mouse Monoclonal Antibody [Clone: FP5]

Supplier: ANTIBODIES.COM
A305038-100
ANTIA305038-100EA 475 EUR
ANTIA305038-100
Anti-Sodium Iodide Symporter Mouse Monoclonal Antibody [Clone: FP5]
Antibodies
Mouse monoclonal [FP5] antibody to Sodium Iodide Symporter for WB, IHC, ICC/IF and antibody microarray with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF, Antibody microarray
  • Recommended dilutions: WB: 1:1000, IHC: 1:1000

Type: Primary
Antigen: Sodium Iodide Symporter
Clonality: Monoclonal
Clone: FP5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR