Anti-PRKAR2B Rabbit Monoclonal Antibody [clone: ARC2272]

Supplier: ANTIBODIES.COM
A305991-100
ANTIA305991-100EA 525 EUR
ANTIA305991-100
Anti-PRKAR2B Rabbit Monoclonal Antibody [clone: ARC2272]
Antibodies
Rabbit monoclonal [ARC2272] antibody to PRKAR2B for WB and IHC with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC
  • Recommended Dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200

Type: Primary
Antigen: PRKAR2B
Clonality: Monoclonal
Clone: ARC2272
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR