Mouse Sorbitol Dehydrogenase ELISA Kit

Supplier: ANTIBODIES.COM

A313534-96
ANTIA313534-96EA 545 EUR
ANTIA313534-96
Mouse Sorbitol Dehydrogenase ELISA Kit
Assays ELISAs

Mouse Sorbitol Dehydrogenase ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Sorbitol Dehydrogenase in serum, plasma, and other biological fluids.


  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 0,75 to 12 mU/ml
  • Sensitivity: 0,028 mU/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%


Mouse Sorbitol Dehydrogenase ELISA kit (A313534) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Sorbitol Dehydrogenase in An antibody specific for Sorbitol Dehydrogenase has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Sorbitol Dehydrogenase present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-Sorbitol Dehydrogenase antibody, which also binds the Sorbitol Dehydrogenase present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Sorbitol Dehydrogenase antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Sorbitol Dehydrogenase antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Sorbitol Dehydrogenase captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Sorbitol Dehydrogenase in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR