Anti-MAPRE1 Rat Monoclonal Antibody [clone: KT51]

Supplier: ANTIBODIES.COM
A35-100
ANTIA35-100EA 445 EUR
ANTIA35-100
Anti-MAPRE1 Rat Monoclonal Antibody [clone: KT51]
Antibodies
Rat monoclonal (KT51) antibody to MAPRE1 for WB, IP and IF with samples derived from mouse, human and ape and hamster.

  • Validated applications: WB, IP, IF
  • Recommended dilutions: WB: 1/500 to 1/10000, IF: 1/200 to 1/1000, IP: 2 µg/ml

Type: Primary
Antigen: MAPRE1
Clonality: Monoclonal
Clone: KT51
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse, Human, Ape and Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR