Mouse Renin ELISA Kit

Supplier: ANTIBODIES.COM

A78731-96
ANTIA78731-96EA 545 EUR
ANTIA78731-96
Mouse Renin ELISA Kit
Assays ELISAs

Mouse Renin ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Renin in serum, plasma, tissue homogenates, and other biological fluids.


  • Ready to use ELISA kit
  • Detection range: 93,75 to 6000 pg/ml
  • Sensitivity: 56,25 pg/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra assay: CV <8%, Inter assay: CV <10%


Mouse Renin ELISA kit (A78731) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Renin in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Renin has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Renin present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Renin Antibody, which binds the captured Renin present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Renin captured in each well. The concentration of Renin can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Order Now

SPECIFICATIONS

Environmentally Preferable
Target protein Renin
Primary antibody reactivity Mouse
Assay Type Sandwich
Assay Principle Quantitative
Host
Format Pre-coated
Assay duration Multiple steps
Sensitivity 56,25 pg/ml
Detection range 93,75 - 6000 pg/ml
Detection method Colorimetric
Cross reactivity Mouse Renin ELISA kit exhibits high specificity and excellent specificity for the detection of mouse Renin. No significant cross-reactivity or interference between Renin and analogues was observed.
Sample type Serum, plasma, tissue homogenates and other biological fluids
Sample volume 100 μl
Time to Results 4 h 30 min
Regulatory status RUO
Storage temperature 4 °C
Shelf life Store for 6 months at 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR