Succinic acid

Supplier: Apollo Scientific
Danger

BI225-1KG BI225-2.5KG BI225-250G
APOSBI225-1KGEA 109 EUR
APOSBI225-1KG APOSBI225-2.5KG APOSBI225-250G
Succinic acid
Succinic acid
Formula: HOOCCH₂CH₂COOH
Boiling Pt: 235 °C
Melting Pt: 184…186 °C
Density: 1,564 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002789
CAS Number: 110-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR