2-Amino-2-methyl-1,3-propanediol 99+%, high purity

Supplier: Apollo Scientific
Warning

BI4545-100G BI4545-25G
APOSBI4545-100GEA 75.4 EUR
APOSBI4545-100G APOSBI4545-25G
2-Amino-2-methyl-1,3-propanediol 99+%, high purity
2-Amino-2-methyl-1,3-propanediol

Buffer component used in SDS - gradient gel electrophoresis. Biological buffer - useful range 7.8-9.7.


  • White crystals
  • Water (by Karl Fischer) <1.0%
  • Forms a clear solution in water.

Formula: C₄H₁₁NO₂
Boiling Pt: 151 °C/10mmHg
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004678
CAS Number: 115-69-5
EINECS: 204-100-7
UN: 3259
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR