HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) free acid ≥99.5%, Ultrapure

Supplier: Apollo Scientific
Warning

BI8181-1KG BI8181-100G BI8181-250G
APOSBI8181-1KGEA 92.8 EUR
APOSBI8181-1KG APOSBI8181-100G APOSBI8181-250G
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) free acid ≥99.5%, Ultrapure
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)

Biological buffer - useful range 6.8 - 8.2


  • Heavy metals: <1 ppm
  • pKa @ 20°C: 7.55 +/- 0.2

Formula: C₈H₁₈N₂O₄S
Melting Pt: 238…240 °C
Storage Temperature: Ambient
MDL Number: MFCD00006158
CAS Number: 7365-45-9
EINECS: 230-907-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR