Amphotericin B

Supplier: Apollo Scientific
Warning

BIA0103-10G BIA0103-1G BIA0103-5G
APOSBIA0103-10GEA 241 EUR
APOSBIA0103-10G APOSBIA0103-1G APOSBIA0103-5G
Amphotericin B
Amphotericin B

A polyene antifungal antibiotic produced by Streptomyces nodosus.

Formula: C₄₇H₇₃NO₁₇
Storage Temperature: Refrigerator
MDL Number: MFCD00877763
CAS Number: 1397-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR