(S)-(+)-2-Amino-1-propanol 99%

Supplier: Apollo Scientific
Danger

Synonyms: L(+)-Alaninol, L-Alaninol, (S)-2-Aminopropan-1-ol, L(+)-2-Amino-1-propanol

BIA0120-100G BIA0120-50G BIA0120-500G
APOSBIA0120-100GEA 29 EUR
APOSBIA0120-100G APOSBIA0120-50G APOSBIA0120-500G
(S)-(+)-2-Amino-1-propanol 99%
(S)-(+)-2-Amino-1-propanol
Formula: C₃H₉NO
Boiling Pt: 174…176 °C
Density: 0,965 g/cm³ (25 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00064412
CAS Number: 2749-11-3
EINECS: 220-388-7
UN: 2735
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR